Learn More
Grainger pH Electrode Epoxy DIN13.5 Round Tip
Catalog No. 502765168
Encompass_Preferred
The SensoRex pH Electrode pH 16Y965 is a gel-filled combination pH electrode with a glass body and epoxy body gel filled reference. It has a pH range of 0-14 pH with a 12 mm diameter x 120 mm length. This electrode has a BNC connector and 3/4" NPT threads. It is compatible with water, wastewater, food processing, and chemical processing applications. The electrode has a built-in temperature sensor and is ideal for continuous online measurement and monitoring of pH.
Catalog No. 50-276-5168 Supplier Grainger Supplier No. 16Y965
The SensoRex pH Electrode pH 16Y965 is a gel-filled combination pH electrode with a glass body and epoxy body gel filled reference. It has a pH range of 0-14 pH with a 12 mm diameter x 120 mm length. This electrode has a BNC connector and 3/4" NPT threads. It is compatible with water, wastewater, food processing, and chemical processing applications. The electrode has a built-in temperature sensor and is ideal for continuous online measurement and monitoring of pH.