Learn More
Grainger pH Electrode Epoxy DIN13.5 Round Tip
Catalog No. 502767673
Encompass_Preferred
The SENSOREX S223CD pH Electrode is a combination pH electrode with a glass body, double junction reference, and BNC connector. It has a pH range of 0-14 pH, temperature range of 0-80°C, and 12/120 mm dimensions. This electrode is designed for laboratory use in aqueous solutions with a fast response time. It requires storage solution and regular calibration for accurate measurements. The S223CD has a 1 year warranty.
Catalog No. 50-276-7673 Supplier Grainger Supplier No. 16Y964
The SENSOREX S223CD pH Electrode is a combination pH electrode with a glass body, double junction reference, and BNC connector. It has a pH range of 0-14 pH, temperature range of 0-80°C, and 12/120 mm dimensions. This electrode is designed for laboratory use in aqueous solutions with a fast response time. It requires storage solution and regular calibration for accurate measurements. The S223CD has a 1 year warranty.