Learn More
Grainger pH Electrode Epoxy DINM/4mm Banana
Catalog No. 502768098
Encompass_Preferred
The SENSOREX S295C is a pH electrode with a glass body and single junction reference. It has a conical tip shape for easy penetration into solids and semisolids. The electrode has a built-in temperature sensor and BNC connector. It has a pH range of 0-14 pH, a temperature range of 0-80°C, and reference electrolyte of KCl gel. The electrode is compatible with the SENSOREX S470 meter. Key features include its rugged design, easy to clean glass, and refillable electrolyte. The S295C is ideal for food, dairy, meat, beverage, and water testing applications.
Catalog No. 50-276-8098 Supplier Grainger Supplier No. 16Y997
The SENSOREX S295C is a pH electrode with a glass body and single junction reference. It has a conical tip shape for easy penetration into solids and semisolids. The electrode has a built-in temperature sensor and BNC connector. It has a pH range of 0-14 pH, a temperature range of 0-80°C, and reference electrolyte of KCl gel. The electrode is compatible with the SENSOREX S470 meter. Key features include its rugged design, easy to clean glass, and refillable electrolyte. The S295C is ideal for food, dairy, meat, beverage, and water testing applications.