Learn More
Grainger pH Electrode Epoxy DINM/C3 Round Tip
Catalog No. 502767980
Encompass_Preferred
The SENSOREX S294C is a pH electrode with a glass body and gel-filled reference. It has a double junction reference with a ceramic pin junction and sleeve junction. The electrode has a conical tip, 12 mm diameter glass body, and BNC connector. It has a pH range of 0-14 pH, temperature range of 0-80°C, and reference electrolyte of 3.8M KCl. The S294C is designed for laboratory, industrial, and municipal water applications. It has a fast response time and stability in fluctuating samples. The double junction reference design minimizes contamination.
Catalog No. 50-276-7980 Supplier Grainger Supplier No. 16Y996
The SENSOREX S294C is a pH electrode with a glass body and gel-filled reference. It has a double junction reference with a ceramic pin junction and sleeve junction. The electrode has a conical tip, 12 mm diameter glass body, and BNC connector. It has a pH range of 0-14 pH, temperature range of 0-80°C, and reference electrolyte of 3.8M KCl. The S294C is designed for laboratory, industrial, and municipal water applications. It has a fast response time and stability in fluctuating samples. The double junction reference design minimizes contamination.