Learn More
Grainger pH Electrode Epoxy STD Round Tip
Catalog No. 502767496
Encompass_Preferred
The Grainger Approved pH Electrode is a combination electrode with a glass body and epoxy body gel filled reference. It has a 12 mm diameter pH bulb and internal Ag/AgCl reference. The electrode has a BNC connector and 1 m cable. It has a polypropylene body and is compatible for use in aqueous solutions from 0-14 pH. The electrode has a temperature range of 0-80°C. Key features include its wide pH range measurement capability and rugged polypropylene body construction.
Catalog No. 50-276-7496 Supplier Grainger Supplier No. 16Y958
The Grainger Approved pH Electrode is a combination electrode with a glass body and epoxy body gel filled reference. It has a 12 mm diameter pH bulb and internal Ag/AgCl reference. The electrode has a BNC connector and 1 m cable. It has a polypropylene body and is compatible for use in aqueous solutions from 0-14 pH. The electrode has a temperature range of 0-80°C. Key features include its wide pH range measurement capability and rugged polypropylene body construction.