Learn More
Hach Company Hach pH Electrode for 8362 Ultrapure Water Sensor (MPN 08362=A=2000)
Catalog No. NC3591258 Shop All Hach Company Products
Encompass
Replacement pH electrode designed for use with the Hach 8362 Ultrapure Water Sensor. It provides a field-replaceable electrode assembly intended for reliable pH measurement in ultrapure water systems.
  • Designed for ultrapure water pH measurement.
  • Compatible with 8362 Ultrapure Water Sensor.
  • Works with a Pt100 temperature sensor for temperature compensation.
  • Field-replaceable electrode assembly for routine maintenance.
  • Connection type: S7.
Catalog No. NC3591258 Supplier Hach Company Supplier No. 08362=A=2000
Replacement pH electrode designed for use with the Hach 8362 Ultrapure Water Sensor. It provides a field-replaceable electrode assembly intended for reliable pH measurement in ultrapure water systems.
  • Designed for ultrapure water pH measurement.
  • Compatible with 8362 Ultrapure Water Sensor.
  • Works with a Pt100 temperature sensor for temperature compensation.
  • Field-replaceable electrode assembly for routine maintenance.
  • Connection type: S7.