Learn More
Grainger pH Electrode Glass BNC Flat Tip 12mm Dia
Catalog No. 502767500
Encompass_Preferred
The Grainger pH Electrode Glass BNC Flat Tip 12mm Dia is a pH electrode with a glass body and flat tip measuring 12mm in diameter. It has a BNC connector and measures pH in aqueous solutions from 0-14 pH. The electrode has a Ag/AgCl reference with a ceramic junction and epoxy body. It has a temperature range of 0-80°C. The electrode provides accurate pH measurements in laboratory applications.
Catalog No. 50-276-7500 Supplier Grainger Supplier No. 16Y982
The Grainger pH Electrode Glass BNC Flat Tip 12mm Dia is a pH electrode with a glass body and flat tip measuring 12mm in diameter. It has a BNC connector and measures pH in aqueous solutions from 0-14 pH. The electrode has a Ag/AgCl reference with a ceramic junction and epoxy body. It has a temperature range of 0-80°C. The electrode provides accurate pH measurements in laboratory applications.