Learn More
Grainger pH Electrode Glass BNC Round Tip
Catalog No. 502768124
Encompass_Preferred
The SensoRex pH Electrode is a combination pH electrode with a glass body and gel-filled reference. It has a double junction reference design for reduced contamination. The electrode has a pH range of 0-14 pH with an accuracy of ±0.02 pH. It has a temperature range of 0-80°C. The electrode has a BNC connector and 1 m cable. It is compatible with most pH meters. The electrode requires storage solution and regular calibration for optimal performance. Overall, this is a high quality, general purpose pH electrode well-suited for most laboratory pH measurements.
Catalog No. 50-276-8124 Supplier Grainger Supplier No. 16Y976
The SensoRex pH Electrode is a combination pH electrode with a glass body and gel-filled reference. It has a double junction reference design for reduced contamination. The electrode has a pH range of 0-14 pH with an accuracy of ±0.02 pH. It has a temperature range of 0-80°C. The electrode has a BNC connector and 1 m cable. It is compatible with most pH meters. The electrode requires storage solution and regular calibration for optimal performance. Overall, this is a high quality, general purpose pH electrode well-suited for most laboratory pH measurements.