Learn More
Grainger pH Electrode Glass BNC Round Tip
Catalog No. 502767671
Encompass_Preferred
The SensoRex pH Electrode pH 16Y981 is a gel-filled pH electrode with a glass body and epoxy body gel filled reference. It has a 12 mm diameter pH bulb and internal temperature sensor. The electrode has a BNC connector and 1 m cable. It has a 3/4" NPT thread mounting and is suitable for water, wastewater and environmental applications. The electrode has a pH range of 0-14 pH, temperature range of 0-80°C, and 12 bar pressure range. It provides accurate pH measurement.
Catalog No. 50-276-7671 Supplier Grainger Supplier No. 16Y981
The SensoRex pH Electrode pH 16Y981 is a gel-filled pH electrode with a glass body and epoxy body gel filled reference. It has a 12 mm diameter pH bulb and internal temperature sensor. The electrode has a BNC connector and 1 m cable. It has a 3/4" NPT thread mounting and is suitable for water, wastewater and environmental applications. The electrode has a pH range of 0-14 pH, temperature range of 0-80°C, and 12 bar pressure range. It provides accurate pH measurement.