Learn More
Grainger pH Electrode Glass BNC Round Tip
Catalog No. 502768111
Encompass_Preferred
The SensoRex pH Electrode from Sensorex is a glass body combination pH electrode designed for general purpose pH measurement in the laboratory. It has a 12 mm diameter glass body, gel filled reference with Ag/AgCl reference wire, ceramic junction, and BNC connector. The electrode has a pH range of 0-14 pH, temperature range of 0-80°C, and impedance of less than 150 MΩ. It requires storage solution and features a quick response time. The electrode can be used for testing water, wastewater, food products, chemical solutions, and more. It is compatible with any pH meter, transmitter, or controller with BNC connector.
Catalog No. 50-276-8111 Supplier Grainger Supplier No. 16Y987
The SensoRex pH Electrode from Sensorex is a glass body combination pH electrode designed for general purpose pH measurement in the laboratory. It has a 12 mm diameter glass body, gel filled reference with Ag/AgCl reference wire, ceramic junction, and BNC connector. The electrode has a pH range of 0-14 pH, temperature range of 0-80°C, and impedance of less than 150 MΩ. It requires storage solution and features a quick response time. The electrode can be used for testing water, wastewater, food products, chemical solutions, and more. It is compatible with any pH meter, transmitter, or controller with BNC connector.