Learn More
Grainger pH Electrode Glass BNC Round Tip
Catalog No. 502768113
Encompass_Preferred
The SensoRex pH Electrode is a combination pH electrode with a glass body and gel-filled reference. It has a double junction reference design for reduced contamination. The electrode has a pH range of 0-14 pH, a temperature range of 0-80°C, and 12 mm diameter x 120 mm length. It has a BNC connector and 1 m cable. The electrode is designed for general purpose pH measurement in liquids. It has a fast response time and is compatible with most pH meters. The replaceable reference design allows for extended electrode life.
Catalog No. 50-276-8113 Supplier Grainger Supplier No. 16Y980
The SensoRex pH Electrode is a combination pH electrode with a glass body and gel-filled reference. It has a double junction reference design for reduced contamination. The electrode has a pH range of 0-14 pH, a temperature range of 0-80°C, and 12 mm diameter x 120 mm length. It has a BNC connector and 1 m cable. The electrode is designed for general purpose pH measurement in liquids. It has a fast response time and is compatible with most pH meters. The replaceable reference design allows for extended electrode life.