Learn More
Sensorex PH ELECTRODE HF RESIST
Catalog No. NC9630787
Encompass_Preferred
Sensorex 12mm flat surface double junction pH electrode with HF resistant glass.
Catalog No. NC9630787 Supplier Sensorex Supplier No. S350CDHF
Sensorex 12mm flat surface double junction pH electrode with HF resistant glass.