Learn More
Cole-Parmer Cole-Parmer pH Electrode Housing, submersible, CPVC, no ATC
Catalog No. 50633493 Shop All Cole-Parmer Products
Encompass_Preferred
These unique holders feature an integrated 316 stainless steel pin ground wire that serves as a temperature compensator and a solution ground. Choose economical CPVC housing or chemical-resistant KYNAR housing. In-line holders have a 1'' NPT(M) connection. Submersible holders have a 3/4'' NPT(F) thread--connect plastic pipe to match your required submersion depth.
Catalog No. 50-633-493 Supplier Cole-Parmer Supplier No. 0572540
These unique holders feature an integrated 316 stainless steel pin ground wire that serves as a temperature compensator and a solution ground. Choose economical CPVC housing or chemical-resistant KYNAR housing. In-line holders have a 1'' NPT(M) connection. Submersible holders have a 3/4'' NPT(F) thread--connect plastic pipe to match your required submersion depth.