Learn More
Grainger pH Electrode Ryton PPS BNC Flat Tip
Catalog No. 502765155
Encompass_Preferred
The SensoRex pH Electrode pH 16Z005 is a gel-filled combination pH electrode with a glass body and epoxy body gel filled reference. It has a 12 mm diameter pH bulb and internal Ag/AgCl reference electrode. The electrode has a BNC connector and 1 m cable. It has a 3M KCl electrolyte solution. The electrode has a pH range of 0-14 pH with a temperature range of 0-80°C. It has a response time of 95% in 1 minute.
Catalog No. 50-276-5155 Supplier Grainger Supplier No. 16Z005
The SensoRex pH Electrode pH 16Z005 is a gel-filled combination pH electrode with a glass body and epoxy body gel filled reference. It has a 12 mm diameter pH bulb and internal Ag/AgCl reference electrode. The electrode has a BNC connector and 1 m cable. It has a 3M KCl electrolyte solution. The electrode has a pH range of 0-14 pH with a temperature range of 0-80°C. It has a response time of 95% in 1 minute.