Learn More
Grainger pH Electrode Ryton PPS BNC Round Tip
Catalog No. 502765154
Encompass_Preferred
The SensoRex pH Electrode pH 16Z001 is a gel-filled combination pH electrode with a glass body and epoxy body gel filled reference. It has a 12 mm diameter pH bulb and internal Ag/AgCl reference electrode. The electrode has a BNC connector and 1 m cable. It has a 3 mol/L KCl electrolyte solution. The electrode has a pH range of 0-14 pH with an accuracy of ±0.02 pH. It has a temperature range of 0-80°C. The response time is ≤10 seconds. This electrode is designed for laboratory, industrial, and municipal water testing applications. It requires storage solution when not in use.
Catalog No. 50-276-5154 Supplier Grainger Supplier No. 16Z002
The SensoRex pH Electrode pH 16Z001 is a gel-filled combination pH electrode with a glass body and epoxy body gel filled reference. It has a 12 mm diameter pH bulb and internal Ag/AgCl reference electrode. The electrode has a BNC connector and 1 m cable. It has a 3 mol/L KCl electrolyte solution. The electrode has a pH range of 0-14 pH with an accuracy of ±0.02 pH. It has a temperature range of 0-80°C. The response time is ≤10 seconds. This electrode is designed for laboratory, industrial, and municipal water testing applications. It requires storage solution when not in use.