Learn More
Grainger pH Electrode Ryton PPS BNC Round Tip
Catalog No. 502768222
Encompass_Preferred
The SENSOREX pH Electrode pH 16Z004 is a pH electrode with a glass body and gel-filled reference. It has a double junction reference with a ceramic junction and sleeve junction. The electrode has a BNC connector and epoxy body. It has a pH range of 0-14 pH with a temperature range of 0-80°C. The electrode has a response time of less than 10 seconds and is compatible with aqueous solutions. It requires storage solution and regular calibration for optimal performance. The electrode measures pH in laboratory samples and processes.
Catalog No. 50-276-8222 Supplier Grainger Supplier No. 16Z004
The SENSOREX pH Electrode pH 16Z004 is a pH electrode with a glass body and gel-filled reference. It has a double junction reference with a ceramic junction and sleeve junction. The electrode has a BNC connector and epoxy body. It has a pH range of 0-14 pH with a temperature range of 0-80°C. The electrode has a response time of less than 10 seconds and is compatible with aqueous solutions. It requires storage solution and regular calibration for optimal performance. The electrode measures pH in laboratory samples and processes.