Learn More
Grainger pH Electrode Ryton PPS BNC Round Tip
Catalog No. 502765148
Encompass_Preferred
The SensoRex pH Electrode pH 16Z001 is a pH electrode with a glass body and epoxy body junction suitable for laboratory use. It has a pH range of 0-14 pH with an accuracy of ±0.02 pH. The electrode has a Ag/AgCl reference with a ceramic junction and uses a BNC connector. It has a temperature range of 0-80°C. The electrode requires a filling solution such as KCl and has a 12 month warranty. The electrode measures 120 mm length x 12 mm diameter.
Catalog No. 50-276-5148 Supplier Grainger Supplier No. 16Z001
The SensoRex pH Electrode pH 16Z001 is a pH electrode with a glass body and epoxy body junction suitable for laboratory use. It has a pH range of 0-14 pH with an accuracy of ±0.02 pH. The electrode has a Ag/AgCl reference with a ceramic junction and uses a BNC connector. It has a temperature range of 0-80°C. The electrode requires a filling solution such as KCl and has a 12 month warranty. The electrode measures 120 mm length x 12 mm diameter.