Learn More
Grainger pH Electrode Ryton PPS BNC/Tinned Leads
Catalog No. 502767641
Encompass_Preferred
The Ryton PPS electrodes and probes on Grainger's website are designed for water testing and measurement. They feature corrosion resistant Ryton polyphenylene sulfide bodies and are available in various styles like pH electrodes, ORP electrodes, conductivity probes and dissolved oxygen probes. The electrodes have replaceable elements and are ideal for use in harsh environments. Overall, these probes provide accurate and reliable measurement for water quality testing applications.
Catalog No. 50-276-7641 Supplier Grainger Supplier No. 16Z011
The Ryton PPS electrodes and probes on Grainger's website are designed for water testing and measurement. They feature corrosion resistant Ryton polyphenylene sulfide bodies and are available in various styles like pH electrodes, ORP electrodes, conductivity probes and dissolved oxygen probes. The electrodes have replaceable elements and are ideal for use in harsh environments. Overall, these probes provide accurate and reliable measurement for water quality testing applications.