Learn More
Grainger pH Electrode Ryton PPS TNR Flat Tip
Catalog No. 502768214
Encompass_Preferred
The S8000CD pH Electrode by Sensorex is a combination pH electrode with a glass body, double junction gel filled reference, and BNC connector. It has a pH range of 0-14 pH, temperature range of 0-100°C, and 12/120 mm dimensions. This electrode is designed for laboratory use in aqueous solutions with a fast response time. It requires storage solution and regular calibration for accurate measurements. The S8000CD has a 1 year warranty.
Catalog No. 50-276-8214 Supplier Grainger Supplier No. 16Z025
The S8000CD pH Electrode by Sensorex is a combination pH electrode with a glass body, double junction gel filled reference, and BNC connector. It has a pH range of 0-14 pH, temperature range of 0-100°C, and 12/120 mm dimensions. This electrode is designed for laboratory use in aqueous solutions with a fast response time. It requires storage solution and regular calibration for accurate measurements. The S8000CD has a 1 year warranty.