Learn More
YSI 3P LifeScience PH ELECTRODE SENTIX 41-3 DIN
Catalog No. 50465218
Encompass_Preferred
Epoxy pH combination electrode with gel electrolyte, integrated temperature sensor, waterproof DIN plug and banana plug, 9.9 ft (3 m) cable.
Catalog No. 50-465-218 Supplier YSI 3P LifeScience Supplier No. 103636
Epoxy pH combination electrode with gel electrolyte, integrated temperature sensor, waterproof DIN plug and banana plug, 9.9 ft (3 m) cable.