Learn More
YSI 3P LifeScience PH ELECTRODE SENTIX 41 DIN
Catalog No. 50465219
Encompass_Preferred
Epoxy pH combination electrode with gel electrolyte, integrated temperature sensor, waterproof DIN plug and banana plug, 3.3 ft (1 m) cable.
Catalog No. 50-465-219 Supplier YSI 3P LifeScience Supplier No. 103635
Epoxy pH combination electrode with gel electrolyte, integrated temperature sensor, waterproof DIN plug and banana plug, 3.3 ft (1 m) cable.