Learn More
YSI 3P LifeScience PH ELECTRODE SENTIX 62
Catalog No. NC9461694
Encompass_Preferred
Glass pH combination electrode with liquid electrolyte, BNC plug. Length 120 mm, 3.3 ft (1 m) cable.
Catalog No. NC9461694 Supplier YSI 3P LifeScience Supplier No. 103641
Glass pH combination electrode with liquid electrolyte, BNC plug. Length 120 mm, 3.3 ft (1 m) cable.