Learn More
Hach Company pH Encap Sens-LCP-2Wire-Convrt
Catalog No. 5000207427 Shop All Hach Company Products
Encompass
pH Encapsulated Sensor-LCP Body-2 Wire-Convertible-FKM/FPM Seals-Stainless Steel Ground Rod-10-ft CableLCP (Liquid Crystal Polymer) Convertible Mounting Style 3m Analog Cable 2-Wire with Built-In Tranmitter General Purpose Glass pH Electrode Stainless Steel Ground Rod
Catalog No. 50-002-07427 Supplier Hach Company Supplier No. 6428P010010000N
pH Encapsulated Sensor-LCP Body-2 Wire-Convertible-FKM/FPM Seals-Stainless Steel Ground Rod-10-ft CableLCP (Liquid Crystal Polymer) Convertible Mounting Style 3m Analog Cable 2-Wire with Built-In Tranmitter General Purpose Glass pH Electrode Stainless Steel Ground Rod