Learn More
Hach Company pH Encap Sens-LCP-2Wire-Convrt
Catalog No. 5000207875 Shop All Hach Company Products
Encompass
pH Encapsulated Sensor-LCP Body-2 Wire-Convertible-FKM/FPM Seals-20 ft CableLCP (Liquid Crystal Polymer) Convertible Mounting Style 6m Analog Cable 2-Wire with Built-In Transmitter General Purpose Glass pH Electrode
Catalog No. 50-002-07875 Supplier Hach Company Supplier No. 6428P020
pH Encapsulated Sensor-LCP Body-2 Wire-Convertible-FKM/FPM Seals-20 ft CableLCP (Liquid Crystal Polymer) Convertible Mounting Style 6m Analog Cable 2-Wire with Built-In Transmitter General Purpose Glass pH Electrode