Learn More
Hach Company pH Encap Sens-LCP-5W-FFKM/FFPM
Catalog No. 5000207871 Shop All Hach Company Products
Encompass
pH Encapsulated Sensor-LCP Body-5 Wire-Convertible-FFKM/FFPM O-Rings-25 ft CableLCP (Liquid Crystal Polymer) Convertible Mounting Style 7 5m Analog Cable 5-Wire with Built-In Preamplifier General Purpose Glass pH Electrode Ceramic/Ceramic Salt Bridge 25 ft Cable
Catalog No. 50-002-07871 Supplier Hach Company Supplier No. 6028P025000010N
pH Encapsulated Sensor-LCP Body-5 Wire-Convertible-FFKM/FFPM O-Rings-25 ft CableLCP (Liquid Crystal Polymer) Convertible Mounting Style 7 5m Analog Cable 5-Wire with Built-In Preamplifier General Purpose Glass pH Electrode Ceramic/Ceramic Salt Bridge 25 ft Cable