Learn More
Hach Company pH Encap Sens-LCP-5W-PreAmp
Catalog No. 5000207416 Shop All Hach Company Products
Encompass
pH Encapsulated Sensor-LCP Body-5 Wire-Convertible-50 ft Cable with Amp Quick DisconnectLCP (Liquid Crystal Polymer) Convertible Mounting Style 15m Analog Cable 5-Wire with Built-In Preamplifier General Purpose Glass pH Electrode
Catalog No. 50-002-07416 Supplier Hach Company Supplier No. 6028P050
pH Encapsulated Sensor-LCP Body-5 Wire-Convertible-50 ft Cable with Amp Quick DisconnectLCP (Liquid Crystal Polymer) Convertible Mounting Style 15m Analog Cable 5-Wire with Built-In Preamplifier General Purpose Glass pH Electrode