Learn More
Hach Company pH Encap Sens-LCP-5W-S-GLS
Catalog No. 5000207582 Shop All Hach Company Products
Encompass
pH Encapsulated Sensor-LCP Body-5 Wire-Convertible- Sin Glass-10 ft CableLCP (Liquid Crystal Polymer) Convertible Mounting Style 3m Analog Cable 5-Wire with Built-In Preamplifier High pH Glass Electrode
Catalog No. 50-002-07582 Supplier Hach Company Supplier No. 6028P210000010N
pH Encapsulated Sensor-LCP Body-5 Wire-Convertible- Sin Glass-10 ft CableLCP (Liquid Crystal Polymer) Convertible Mounting Style 3m Analog Cable 5-Wire with Built-In Preamplifier High pH Glass Electrode