Learn More
Hach Company pH Encap Sens-LCP-Convert-5Wr
Catalog No. 5000208094 Shop All Hach Company Products
Encompass
pH Encapsulated Sensor-LCP Body-5 Wire-Convertible-Low pH Hin Glass-10 ft CableLCP (Liquid Crystal Polymer) Convertible Mounting Style 3m Analog Cable 5-Wire with Built-In Preamplifier Low pH Glass Electrode
Catalog No. 50-002-08094 Supplier Hach Company Supplier No. 6028P6
pH Encapsulated Sensor-LCP Body-5 Wire-Convertible-Low pH Hin Glass-10 ft CableLCP (Liquid Crystal Polymer) Convertible Mounting Style 3m Analog Cable 5-Wire with Built-In Preamplifier Low pH Glass Electrode