Learn More
Hach Company pH Encap Sens-LCP-Convert-5Wr
Catalog No. 5000207825 Shop All Hach Company Products
Encompass
pH Encapsulated Sensor-LCP Body-5 Wire-Convertible-Low Conductivity-20 ft CableLCP (Liquid Crystal Polymer) Convertible Mounting Style 6m Analog Cable 5-Wire with Built-In Preamplifier pH Glass Electrode for Low Conductivity Solutions
Catalog No. 50-002-07825 Supplier Hach Company Supplier No. 6028P420
pH Encapsulated Sensor-LCP Body-5 Wire-Convertible-Low Conductivity-20 ft CableLCP (Liquid Crystal Polymer) Convertible Mounting Style 6m Analog Cable 5-Wire with Built-In Preamplifier pH Glass Electrode for Low Conductivity Solutions