Learn More
Hach Company pH Encap Sens-LCP-Flow-Thru-5
Catalog No. 5000207430 Shop All Hach Company Products
Encompass
pH Encapsulated Sensor-LCP Body-5 Wire-Flow-Thru Mount-with Amphenol Connector-10 ft CableLCP (Liquid Crystal Polymer) Union Mounting Style 3m Analog Cable 5-Wire with Built-In Preamplifier General Purpose Glass pH Electrode Circular MIL Connector - 7 Pin
Catalog No. 50-002-07430 Supplier Hach Company Supplier No. 6058P01000A000N
pH Encapsulated Sensor-LCP Body-5 Wire-Flow-Thru Mount-with Amphenol Connector-10 ft CableLCP (Liquid Crystal Polymer) Union Mounting Style 3m Analog Cable 5-Wire with Built-In Preamplifier General Purpose Glass pH Electrode Circular MIL Connector - 7 Pin