Learn More
Hach Company pH Encap Sens-PPS-5 Wr-Convert
Catalog No. 5000207805 Shop All Hach Company Products
Encompass
pH Encapsulated Sensor-PPS Body-5 Wire-Convertible-Antimony Electrode Material-10-ft CablePPS Convertible Mounting Style 3m Analog Cable 5-Wire with Built-In Preamplifier Antimony pH Electrode
Catalog No. 50-002-07805 Supplier Hach Company Supplier No. 6022P1
pH Encapsulated Sensor-PPS Body-5 Wire-Convertible-Antimony Electrode Material-10-ft CablePPS Convertible Mounting Style 3m Analog Cable 5-Wire with Built-In Preamplifier Antimony pH Electrode