Learn More
Hach Company pH Encapsulated Sensor, LCP Body, 2-Wire Convertible, FKM/FPM Seals, 50 ft Cable
Catalog No. 5000207848 Shop All Hach Company Products
Encompass
Encapsulated pH sensor designed for reliable online pH monitoring in industrial and water-treatment applications. It features an LCP-encapsulated body, a general-purpose glass electrode, convertible mounting, and a factory-attached 15 m (50 ft) cable. An optional built-in two-wire transmitter provides a 4-20 mA output and a built-in preamplifier for long cable runs.
  • Body constructed from liquid crystal polymer for chemical resistance.
  • Convertible mounting with 1.5 in NPT threads for immersion or inline use.
  • Factory-attached 15 m (50 ft) 2-conductor cable for easy installation.
  • Optional two-wire transmitter provides 4-20 mA output.
  • Built-in preamplifier supports analyzer placement up to 3,000 ft.
  • FKM/FPM seals and PVDF junction for improved durability.
  • General-purpose glass electrode suitable for routine pH measurement.
Catalog No. 50-002-07848 Supplier Hach Company Supplier No. 6428P050
Encapsulated pH sensor designed for reliable online pH monitoring in industrial and water-treatment applications. It features an LCP-encapsulated body, a general-purpose glass electrode, convertible mounting, and a factory-attached 15 m (50 ft) cable. An optional built-in two-wire transmitter provides a 4-20 mA output and a built-in preamplifier for long cable runs.
  • Body constructed from liquid crystal polymer for chemical resistance.
  • Convertible mounting with 1.5 in NPT threads for immersion or inline use.
  • Factory-attached 15 m (50 ft) 2-conductor cable for easy installation.
  • Optional two-wire transmitter provides 4-20 mA output.
  • Built-in preamplifier supports analyzer placement up to 3,000 ft.
  • FKM/FPM seals and PVDF junction for improved durability.
  • General-purpose glass electrode suitable for routine pH measurement.