Learn More
Hach Company pH Encapsulated Sensor
Catalog No. 5000207706 Shop All Hach Company Products
Encompass
pH Encapsulated Sensor-LCP Body-5 Wire-Flow-Thru Mount-H Glass-Gel Filled Standard-Ceramic/Ceramic Saltbridge-10 ft Cable LCP (Liquid Crystal Polymer) Union Mounting Style 3m Analog Cable 5-Wire with Built-In Preamplifier Low pH Glass Electrode Gel Filled Standard Ceramic/Ceramic Salt Bridge
Catalog No. 50-002-07706 Supplier Hach Company Supplier No. 6058P610F00010N
pH Encapsulated Sensor-LCP Body-5 Wire-Flow-Thru Mount-H Glass-Gel Filled Standard-Ceramic/Ceramic Saltbridge-10 ft Cable LCP (Liquid Crystal Polymer) Union Mounting Style 3m Analog Cable 5-Wire with Built-In Preamplifier Low pH Glass Electrode Gel Filled Standard Ceramic/Ceramic Salt Bridge