Learn More
Hach Company pH Encapsulated Sensor
Catalog No. 5000207823 Shop All Hach Company Products
Encompass
pH Encapsulated Sensor-LCP Body-5 Wire-Union Mount-Antimony Electrode Material-10 ft Cable LCP (Liquid Crystal Polymer) Union Mounting Style 3m Analog Cable 5-Wire with Built-In Preamplifier Antimony pH Electrode
Catalog No. 50-002-07823 Supplier Hach Company Supplier No. 6058P1
pH Encapsulated Sensor-LCP Body-5 Wire-Union Mount-Antimony Electrode Material-10 ft Cable LCP (Liquid Crystal Polymer) Union Mounting Style 3m Analog Cable 5-Wire with Built-In Preamplifier Antimony pH Electrode