Learn More
Hach Company pH Encapsulated Sensor
Catalog No. 5000208096 Shop All Hach Company Products
Encompass
pH Encapsulated Sensor-LCP Body-5 Wire-Union Mount-High Purity Water Glass-Low Conductivity-10 ft Cable LCP (Liquid Crystal Polymer) Union Mounting Style 3m Analog Cable 5-Wire with Built-In Preamplifier pH Glass Electrode for Low Conductivity Solutions
Catalog No. 50-002-08096 Supplier Hach Company Supplier No. 6058P4
pH Encapsulated Sensor-LCP Body-5 Wire-Union Mount-High Purity Water Glass-Low Conductivity-10 ft Cable LCP (Liquid Crystal Polymer) Union Mounting Style 3m Analog Cable 5-Wire with Built-In Preamplifier pH Glass Electrode for Low Conductivity Solutions