Learn More
Broadley James Corporation PH FERM PROBE (325MM)
Catalog No. NC9894751
Encompass
Model F-635 pH FermProbe with Standard Metric Cap,Electrode Length:325 mm,Specifications:0 - 13 pH range -- Steam sterilizable to 135 deg.C -- 150 psig max. pressure -- Double junction,Ag-AgCl reference system
Catalog No. NC9894751 Supplier Broadley James Corporation Supplier No. F635B325DH
Model F-635 pH FermProbe with Standard Metric Cap,Electrode Length:325 mm,Specifications:0 - 13 pH range -- Steam sterilizable to 135 deg.C -- 150 psig max. pressure -- Double junction,Ag-AgCl reference system