Learn More
Broadley James Corporation F-695 Autoclavable pH Electrode
Catalog No. NC0548927
Encompass
The F-695 is similar to the F-635 and varies only in the connector type. Suitable for either benchtop or production vessels. Steam sterilizable and autoclavable.FeaturesMetric threaded cap is compatible with nearly all European designed bioprocess electrode housings.Steam sterilizable and autoclavable.First choice for use with small autoclavable vessels.Available in variety of lengths for different insertion length requirements and applications.Specifications 0- 13 pH rangeSteam sterilizable to 135° C150 psig max. pressureDouble junction, Ag-AgCl reference systemMetric Pg13.5 threaded disconnect cap
Catalog No. NC0548927 Supplier Broadley James Corporation Supplier No. F695B120DK
The F-695 is similar to the F-635 and varies only in the connector type. Suitable for either benchtop or production vessels. Steam sterilizable and autoclavable.FeaturesMetric threaded cap is compatible with nearly all European designed bioprocess electrode housings.Steam sterilizable and autoclavable.First choice for use with small autoclavable vessels.Available in variety of lengths for different insertion length requirements and applications.Specifications 0- 13 pH rangeSteam sterilizable to 135° C150 psig max. pressureDouble junction, Ag-AgCl reference systemMetric Pg13.5 threaded disconnect cap