Learn More
Broadley James Corporation F-815 Sterilizable pH Electrode
Catalog No. NC1431554
Encompass
The new F-815 offers the same form, fit and function as the F-615 with improved performance and USP Class VI materials. The unique construction methods used by Broadley-James allows this pH probe to exhibit virtually zero drift after steam sterilization (SIP). Use with these Housing Models: 320, 330, 326, 336, 350, 357, 370.
Catalog No. NC1431554 Supplier Broadley James Corporation Supplier No. F815B130DH
The new F-815 offers the same form, fit and function as the F-615 with improved performance and USP Class VI materials. The unique construction methods used by Broadley-James allows this pH probe to exhibit virtually zero drift after steam sterilization (SIP). Use with these Housing Models: 320, 330, 326, 336, 350, 357, 370.