Learn More
CTL Scientific Supply Corp PH-FIX 4.0-7.0,100 TEST STRIPS
SDP
Supplier:  CTL Scientific Supply Corp 92137
Encompass_Preferred
PH-FIX 4.0 - 7.0 are non-bleeding indicator strips. Gradation of scale: 4.0 - 4.4 - 4.7 - 5.0 - 5.3 - 5.5 - 5.8 - 6.1 - 6.5 - 7.0, box of 100 strips
Catalog No. 50-117-3328