Learn More
Macherey-Nagel pH test strips, pH‑Fix 0–14, fixed indicator, 100/pk
Catalog No. NC0963658
Encompass_Preferred
pH‑Fix strips with chemically bound indicators are the bench mark for pH testing with test strips. The fixation prevents bleeding and therefore contamination of the sample. pH‑Fix 0–14 is also suitable for automatic evaluation with QUANTOFIX Relax.
Catalog No. NC0963658 Supplier Macherey-Nagel Supplier No. 92110
pH‑Fix strips with chemically bound indicators are the bench mark for pH testing with test strips. The fixation prevents bleeding and therefore contamination of the sample. pH‑Fix 0–14 is also suitable for automatic evaluation with QUANTOFIX Relax.