Learn More
CTL Scientific Supply Corp PH-FIX ST. 1.7-3.8,CS/10BX/100
SDP
Supplier:  CTL Scientific Supply Corp 9219010
Encompass_Preferred
PH-FIX 1.7-3.8 TEST PAPER are non-bleeding indicator strips. Gradation of scale: 1.7 - 2.0 - 2.3 - 2.6 - 2.9 - 3.2 - 3.5 - 3.8 (case of 10 boxes of 100 strips each)
Catalog No. 50-117-3347