Learn More
CTL Scientific Supply Corp PH-FIX STR. .3-2.3,CS/10BX/100
SDP
Supplier:  CTL Scientific Supply Corp 9218010
Encompass_Preferred
PH-FIX 0.3-2.3 TEST PAPER are non-bleeding indicator strips. Gradation of scale: 0.3 - 0.7 - 1.0 - 1.3 - 1.6 - 1.9 - 2.3 (case of 10 boxes of 100 strips each)
Catalog No. 50-117-3346