Learn More
CTL Scientific Supply Corp PH-FIX STRIPS 0-6,CS/10BX/100
SDP
Supplier:  CTL Scientific Supply Corp 9211510
Encompass_Preferred
PH-FIX 0.0-6.0 TEST PAPER are non-bleeding indicator strips. Gradation of scale: 0 - 0.5 - 1.0 - 1.5 - 2.0 - 2.5 - 3.0 - 3.5 - 4.0 - 4.5 - 5.0 - 5.5 - 6.0 (case of 10 boxes of 100 strips each)
Catalog No. 50-117-3341