Learn More
Hach Company Intellical™ PHC201 Laboratory General Purposes Gel Filled pH Electrode, 3 m Cable
Catalog No. 501998077 Shop All Hach Company Products
Encompass
Intellical™ PHC201 is a digital combination pH electrode with built-in temperature sensor. The PHC201 has a non-refillable semi-liquid gel-filled ceramic pin reference junction. This laboratory pH electrode is shockproff with its epoxy plastic body protecting down to the glass bulb sensing element. The electrode is available with a 3 or 1 m cable. The PHC201 pH electrode is recommended for general aqueous applications. PHC201 is not suitable for use with organic solvents, low ionic strength, wastewater, high solids or high alkaline sample types.
Catalog No. 50-199-8077 Supplier Hach Company Supplier No. PHC20103
Add to Cart
Add to Cart
Intellical™ PHC201 is a digital combination pH electrode with built-in temperature sensor. The PHC201 has a non-refillable semi-liquid gel-filled ceramic pin reference junction. This laboratory pH electrode is shockproff with its epoxy plastic body protecting down to the glass bulb sensing element. The electrode is available with a 3 or 1 m cable. The PHC201 pH electrode is recommended for general aqueous applications. PHC201 is not suitable for use with organic solvents, low ionic strength, wastewater, high solids or high alkaline sample types.