Learn More
Hach Company Intellical™ PHC101 Field Low Maintenance Gel Filled pH Electrode, 15 m Cable
Catalog No. 501998073 Shop All Hach Company Products
Encompass
Intellical™ PHC101 is a digital combination pH electrode with built-in temperature sensor. PHC101 has low maintenance needs thanks to a non-refillable gel-filled single open reference junction. This Rugged PHC101 version is built with a stainless steel body ensuring sufficient weight to the probe for an easy outside handling, the pH sensing part is protected by a shroud, and the connection is ensured by strong and visible yellow coloured cable onto which you can clip depth markers (optional accessories), making the PHC101 rugged models specially designed for field use. PHC101 Rugged is available with a 5, 10, 15, or 30 meters cable. The PHC101 Rugged is ideal for measuring pH in all types of external environment such as river, surface and ground waters, ponds, lakes, sea, wastewater plant, source water, drinking water reservoir for water quality, environmental and treatment process purposes.
Catalog No. 50-199-8073 Supplier Hach Company Supplier No. PHC10115
Intellical™ PHC101 is a digital combination pH electrode with built-in temperature sensor. PHC101 has low maintenance needs thanks to a non-refillable gel-filled single open reference junction. This Rugged PHC101 version is built with a stainless steel body ensuring sufficient weight to the probe for an easy outside handling, the pH sensing part is protected by a shroud, and the connection is ensured by strong and visible yellow coloured cable onto which you can clip depth markers (optional accessories), making the PHC101 rugged models specially designed for field use. PHC101 Rugged is available with a 5, 10, 15, or 30 meters cable. The PHC101 Rugged is ideal for measuring pH in all types of external environment such as river, surface and ground waters, ponds, lakes, sea, wastewater plant, source water, drinking water reservoir for water quality, environmental and treatment process purposes.