Learn More
YMC America, Inc Ph guard 120A 5um 10x2.1mm
SDP
Catalog No. 5000878736
Encompass
Phenyl columns provide unique selectivity when compared to aliphatic straight chain reversed phase materials such as C18 C8 and C4 YMC-Pack Phenyl is a monomerically-bonded phenyl and is fully endcapped Typical applications include aromatic compounds phenols fullerenes sweeteners
Catalog No. 50-008-78736 Supplier YMC America, Inc Supplier No. PH12S0501Q1GC
Phenyl columns provide unique selectivity when compared to aliphatic straight chain reversed phase materials such as C18 C8 and C4 YMC-Pack Phenyl is a monomerically-bonded phenyl and is fully endcapped Typical applications include aromatic compounds phenols fullerenes sweeteners