Learn More
YMC America, Inc Ph-HG col 120A 10um 250x6mm
SDP
Catalog No. 5000880677
Encompass_Preferred
Phenyl columns provide unique selectivity when compared to aliphatic straight chain reversed phase materials such as C18 C8 and C4 YMC-Pack Phenyl is a monomerically-bonded phenyl and is fully endcapped Typical applications include aromatic compounds phenols fullerenes sweeteners
Catalog No. 50-008-80677 Supplier YMC America, Inc Supplier No. PHG12S112506WT
Add to Cart
Add to Cart
Phenyl columns provide unique selectivity when compared to aliphatic straight chain reversed phase materials such as C18 C8 and C4 YMC-Pack Phenyl is a monomerically-bonded phenyl and is fully endcapped Typical applications include aromatic compounds phenols fullerenes sweeteners