Learn More
YMC America, Inc Ph-HG col 120A 15um 250x30mm
SDP
Catalog No. 5000881633
Encompass
Phenyl columns provide unique selectivity when compared to aliphatic straight chain reversed phase materials such as C18 C8 and C4 YMC-Pack Phenyl is a monomerically-bonded phenyl and is fully endcapped Typical applications include aromatic compounds phenols fullerenes sweeteners
Catalog No. 50-008-81633 Supplier YMC America, Inc Supplier No. PHG12S162530WT
Phenyl columns provide unique selectivity when compared to aliphatic straight chain reversed phase materials such as C18 C8 and C4 YMC-Pack Phenyl is a monomerically-bonded phenyl and is fully endcapped Typical applications include aromatic compounds phenols fullerenes sweeteners