Learn More
Electron Microscopy Sciences pH Indicator Papers - Test Papers, 0.0 - 14, 100/bx
SDP
Catalog No. 5027834
Encompass_Preferred
The indicator dyes of pH-Fix sticks are substantially bound to the cellulose fibers and as a result the possibility of the color bleeding, even in strongly basic solutions, is avoided. The dyes provide a sharp differentiation between the individual pH values and a clear and easy comparison to the color scale. Measurement of pH value is possible even in unbuffered or very weakly buffered solutions, since the sticks can be left in the solution until the final color change is completed. Test samples are not contaminated by the indicator dyes.
Gradations: 1
Catalog No. 50-278-34 Supplier Electron Microscopy Sciences Supplier No. 6551010
The indicator dyes of pH-Fix sticks are substantially bound to the cellulose fibers and as a result the possibility of the color bleeding, even in strongly basic solutions, is avoided. The dyes provide a sharp differentiation between the individual pH values and a clear and easy comparison to the color scale. Measurement of pH value is possible even in unbuffered or very weakly buffered solutions, since the sticks can be left in the solution until the final color change is completed. Test samples are not contaminated by the indicator dyes.
Gradations: 1