Learn More
Med Vet International pH Indicator Paper w/pH range 4-9, J0892, 15ft, 1 Roll
Catalog No. 5000582036
Encompass_Preferred
pH Indicator Paper. Knowledge of body solution or fluid pH (i.e., urine, ruminal con- tents and abdominal tap) is required frequently in veterinary medicine.pH papers provide a fast, simple, and economical method of pH measurements. For most solutions, it is only necessary to dip a short strip of test paper in the solution for a second or two. Then, shake off the excess fluid, match the color of the wet test paper with its color chart, and read off the value.pH Paper Roll in Plastic Dispenser. Test strips on a continuous roll, tear off what you need. pH range 4-9. 15'.
Catalog No. 50-005-82036 Supplier Med Vet International Supplier No. JOR892
pH Indicator Paper. Knowledge of body solution or fluid pH (i.e., urine, ruminal con- tents and abdominal tap) is required frequently in veterinary medicine.pH papers provide a fast, simple, and economical method of pH measurements. For most solutions, it is only necessary to dip a short strip of test paper in the solution for a second or two. Then, shake off the excess fluid, match the color of the wet test paper with its color chart, and read off the value.pH Paper Roll in Plastic Dispenser. Test strips on a continuous roll, tear off what you need. pH range 4-9. 15'.